3-Amino-4-chloro-1H-indazole
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Chloro-1H-indazol-3-amine
OR9905-25G
OR9905-5G
APOSOR9905-25GEA
8270
CZK
APOSOR9905-25G
APOSOR9905-5G
3-Amino-4-chloro-1H-indazole
3-Amino-4-chloro-1H-indazole
Vzorec:
C₇H₆ClN₃ MW: 167,60 g/mol Bod tání: 159…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00663139 CAS číslo: 20925-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...