Tisk…

Imidazo[1,2-a]pyridine-2-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR9880-1G OR9880-100MG OR9880-250MG
APOSOR9880-1GEA 953 CZK
APOSOR9880-1G APOSOR9880-100MG APOSOR9880-250MG
Imidazo[1,2-a]pyridine-2-carboxylic acid
Imidazo[1,2-a]pyridine-2-carboxylic acid
Vzorec: C₈H₆N₂O₂
MW: 162,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03419462
CAS číslo: 64951-08-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR