Ethyl-6-oxo-2-phenyl-1,6-dihydro-5-pyrimidinecarboxylate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Ethyl 1,6-dihydro-6-oxo-2-phenylpyrimidine-5-carboxylate
OR9847-1G
OR9847-5G
APOSOR9847-1GEA
870
CZK
APOSOR9847-1G
APOSOR9847-5G
Ethyl-6-oxo-2-phenyl-1,6-dihydro-5-pyrimidinecarboxylate
Ethyl-6-oxo-2-phenyl-1,6-dihydro-5-pyrimidinecarboxylate
Vzorec:
C₁₃H₁₂N₂O₃ MW: 244,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00695462 CAS číslo: 55613-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...