4-Amino-1-boc-piperidine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
tert-Butyl 4-Amino-1-piperidinecarboxylate, 4-Amino-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-aminopiperidine-1-carboxylate, N-Boc-4-piperidineamine, tert-Butyl 4-amino-1-piperidinecarboxylate, 4-Amino-1-Boc-piperidine, 4-Amino-1-piperidinecarboxylic acid tert-Butyl Ester
OR9845-100G
OR9845-25G
OR9845-5G
APOSOR9845-100GEA
2070
CZK
APOSOR9845-100G
APOSOR9845-25G
APOSOR9845-5G
4-Amino-1-boc-piperidine 97%
4-Amino-1-boc-piperidine
Vzorec:
C₁₀H₂₀N₂O₂ Bod varu: 80 °C/0,037mmHg Bod tání: 44…48 °C Storage Temperature: Ambient |
MDL Number:
MFCD01076201 CAS číslo: 87120-72-7 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...