Tisk…

4-(5-Butyl-1,3,4-oxadiazol-2-yl)phenylamine

Dodavatel: Apollo Scientific

OR9835-1G OR9835-5G
APOSOR9835-1GEA 4630 CZK
APOSOR9835-1G APOSOR9835-5G
4-(5-Butyl-1,3,4-oxadiazol-2-yl)phenylamine
4-(5-Butyl-1,3,4-oxadiazol-2-yl)phenylamine
Vzorec: C₁₂H₁₅N₃O
Bod tání: 119…121 °C
Storage Temperature: Ambient
MDL Number: MFCD06797387
CAS číslo: 100933-82-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR