3-Isopropoxybenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR9794-1G
OR9794-5G
APOSOR9794-1GEA
3410
CZK
APOSOR9794-1G
APOSOR9794-5G
3-Isopropoxybenzoic acid
3-Isopropoxybenzoic acid
Vzorec:
C₁₀H₁₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD01922170 CAS číslo: 60772-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...