5-Amino-2-chlorobenzoic acid 98%
Dodavatel: Apollo Scientific
OR9778-100G
APOSOR9778-100GEA
2730
CZK
APOSOR9778-100G
5-Amino-2-chlorobenzoic acid 98%
5-Amino-2-chlorobenzoic acid
Vzorec:
C₇H₆ClNO₂ MW: 171,58 g/mol Bod tání: 184…188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007845 CAS číslo: 89-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...