Tisk…

Ethyl-7-methyl-2-phenylimidazo[1,2-a]pyridine-3-carboxylate

Dodavatel: Apollo Scientific
Varování

OR9777-250MG OR9777-50MG
APOSOR9777-250MGEA 5290 CZK
APOSOR9777-250MG APOSOR9777-50MG
Ethyl-7-methyl-2-phenylimidazo[1,2-a]pyridine-3-carboxylate
Ethyl-7-methyl-2-phenylimidazo[1,2-a]pyridine-3-carboxylate
Vzorec: C₁₇H₁₆N₂O₂
Storage Temperature: Ambient
CAS číslo: 137997-34-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR