Tisk…

4-Ethylphenylhydrazine hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Ethylphenylhydrazine Hydrochloride

OR9704-25G
APOSOR9704-25GEA 4470 CZK
APOSOR9704-25G
4-Ethylphenylhydrazine hydrochloride
4-Ethylphenylhydrazine hydrochloride
Vzorec: C₈H₁₃ClN₂
MW: 172,6552 g/mol
Bod tání: 232…234 °C
Storage Temperature: Ambient
MDL Number: MFCD01861267
CAS číslo: 53661-18-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR