Tisk…

1-Propylpiperazine dihydrobromide 98%

Dodavatel: Apollo Scientific

OR968321-25G
APOSOR968321-25GEA 2650 CZK
APOSOR968321-25G
1-Propylpiperazine dihydrobromide 98%
1-Propylpiperazine dihydrobromide
Vzorec: C₇H₁₆N₂
MW: 290,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051992
CAS číslo: 64262-23-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR