N-(6-Hydroxypyridin-2-yl)-2,2-dimethylpropionamide 96%
Dodavatel: Apollo Scientific
OR965882-1G
OR965882-250MG
APOSOR965882-1GEA
12830
CZK
APOSOR965882-1G
APOSOR965882-250MG
N-(6-Hydroxypyridin-2-yl)-2,2-dimethylpropionamide 96%
N-(6-Hydroxypyridin-2-yl)-2,2-dimethylpropionamide
Vzorec:
C₁₀H₁₄N₂O₂ MW: 194,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07366716 CAS číslo: 824429-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...