Tisk…

2-(3-Chlorophenyl)ethanol 98%

Dodavatel: Apollo Scientific

Synonyms: 3-Chlorophenethyl alcohol

OR965862-100G OR965862-25G
APOSOR965862-100GEA 6050 CZK
APOSOR965862-100G APOSOR965862-25G
2-(3-Chlorophenyl)ethanol 98%
2-(3-Chlorophenyl)ethanol
Vzorec: C₈H₉ClO
MW: 156,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002892
CAS číslo: 5182-44-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR