Tisk…

6-Chloro-2-methoxypyrimidin-4-amine 95%

Dodavatel: Apollo Scientific

Synonyms: 4-Amino-6-chloro-2-methoxypyrimidine

OR965821-1G OR965821-250MG OR965821-5G
APOSOR965821-1GEA 4070 CZK
APOSOR965821-1G APOSOR965821-250MG APOSOR965821-5G
6-Chloro-2-methoxypyrimidin-4-amine 95%
6-Chloro-2-methoxypyrimidin-4-amine
Vzorec: C₅H₆ClN₃O
MW: 159,5700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11053596
CAS číslo: 3286-55-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR