Tisk…

4-Pyridin-4-ylpyrimidin-2-amine 95%

Dodavatel: Apollo Scientific

OR965736-1G OR965736-5G
APOSOR965736-1GEA 4770 CZK
APOSOR965736-1G APOSOR965736-5G
4-Pyridin-4-ylpyrimidin-2-amine 95%
4-Pyridin-4-ylpyrimidin-2-amine
Vzorec: C₉H₈N₄
MW: 172,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03305477
CAS číslo: 66521-70-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR