4-(5-Oxazolyl)benzylamine 95%
Dodavatel: Apollo Scientific
OR965662-100MG
OR965662-1G
OR965662-250MG
APOSOR965662-100MGEA
4140
CZK
APOSOR965662-100MG
APOSOR965662-1G
APOSOR965662-250MG
4-(5-Oxazolyl)benzylamine 95%
4-(5-Oxazolyl)benzylamine
Vzorec:
C₁₀H₁₀N₂O Storage Temperature: Ambient |
MDL Number:
MFCD06213310 CAS číslo: 672324-91-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...