Tisk…

3-(Cyanomethyl)benzoic acid 96%

Dodavatel: Apollo Scientific

OR965652-1G OR965652-250MG
APOSOR965652-1GEA 1290 CZK
APOSOR965652-1G APOSOR965652-250MG
3-(Cyanomethyl)benzoic acid 96%
3-(Cyanomethyl)benzoic acid
Vzorec: C9H7NO2
MW: 161,1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08234486
CAS číslo: 5689-33-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR