Tisk…

5-Methyl-1,3,4-oxadiazole-2-thiol 95%

Dodavatel: Apollo Scientific

OR965582-1G
APOSOR965582-1GEA 3970 CZK
APOSOR965582-1G
5-Methyl-1,3,4-oxadiazole-2-thiol 95%
5-Methyl-1,3,4-oxadiazole-2-thiol
Vzorec: C₃H₄N₂OS
Storage Temperature: Ambient
MDL Number: MFCD03161246
CAS číslo: 31130-17-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR