Tisk…

2-Pyrrolidin-1-ylbenzaldehyde 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-(1-Pyrrolidinyl)benzaldehyde

OR9655-1G OR9655-100MG OR9655-5G
APOSOR9655-1GEA 2700 CZK
APOSOR9655-1G APOSOR9655-100MG APOSOR9655-5G
2-Pyrrolidin-1-ylbenzaldehyde 97%
2-Pyrrolidin-1-ylbenzaldehyde
Vzorec: C₁₁H₁₃NO
MW: 175,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03419322
CAS číslo: 58028-74-3
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR