Tisk…

(E)-2-Chloro-5-methylnicotinaldehyde oxime 96%

Dodavatel: Apollo Scientific

OR965476-1G OR965476-5G
APOSOR965476-1GEA 6220 CZK
APOSOR965476-1G APOSOR965476-5G
(E)-2-Chloro-5-methylnicotinaldehyde oxime 96%
(E)-2-Chloro-5-methylnicotinaldehyde oxime
Vzorec: C₇H₇ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD13563057
CAS číslo: 1203500-13-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR