Tisk…

2-(3,4-Dihydroisoquinolin-2(1H)-yl)ethanamine 95%

Dodavatel: Apollo Scientific

OR965459-1G OR965459-5G
APOSOR965459-1GEA 7670 CZK
APOSOR965459-1G APOSOR965459-5G
2-(3,4-Dihydroisoquinolin-2(1H)-yl)ethanamine 95%
2-(3,4-Dihydroisoquinolin-2(1H)-yl)ethanamine
Vzorec: C₁₁H₁₆N₂
Storage Temperature: Ambient
MDL Number: MFCD01717314
CAS číslo: 53356-51-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR