Tisk…

4-(2-Aminoethyl)benzonitrile hydrochloride 97%

Dodavatel: Apollo Scientific

OR965409-1G OR965409-5G
APOSOR965409-1GEA 4140 CZK
APOSOR965409-1G APOSOR965409-5G
4-(2-Aminoethyl)benzonitrile hydrochloride 97%
4-(2-Aminoethyl)benzonitrile hydrochloride
Vzorec: C₉H₁₁ClN₂
Storage Temperature: Ambient
CAS číslo: 167762-80-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR