1-(1-Methyl-1H-pyrazol-5-yl)ethanone 95%
Dodavatel: Apollo Scientific
OR965394-1G
OR965394-250MG
APOSOR965394-1GEA
15310
CZK
APOSOR965394-1G
APOSOR965394-250MG
1-(1-Methyl-1H-pyrazol-5-yl)ethanone 95%
1-(1-Methyl-1H-pyrazol-5-yl)ethanone
Vzorec:
C₆H₈N₂O Storage Temperature: Ambient |
MDL Number:
MFCD04968784 CAS číslo: 137890-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...