Tisk…

Ethyl 3-Cyclohexyl-3-oxopropionate 95%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 3-Cyclohexyl-3-oxopropanoate, 3-Cyclohexyl-3-oxopropionic acid Ethyl ester

OR965283-5G
APOSOR965283-5GEA 2780 CZK
APOSOR965283-5G
Ethyl 3-Cyclohexyl-3-oxopropionate 95%
Ethyl 3-Cyclohexyl-3-oxopropionate
Vzorec: C₁₁H₁₈O₃
MW: 198.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115574
CAS číslo: 15971-92-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR