3-(3-Methoxyphenoxy)propylamine 95%
Dodavatel: Apollo Scientific
OR965267-1G
OR965267-250MG
APOSOR965267-1GEA
10350
CZK
APOSOR965267-1G
APOSOR965267-250MG
3-(3-Methoxyphenoxy)propylamine 95%
3-(3-Methoxyphenoxy)propylamine
Vzorec:
C₁₀H₁₅NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD07788402 CAS číslo: 6451-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...