Tisk…

3-Ethylpyridin-2-amine 96%

Dodavatel: Apollo Scientific

OR965045-10G OR965045-1G OR965045-5G
APOSOR965045-10GEA 24600 CZK
APOSOR965045-10G APOSOR965045-1G APOSOR965045-5G
3-Ethylpyridin-2-amine 96%
3-Ethylpyridin-2-amine
Vzorec: C₇H₁₀N₂
Storage Temperature: Ambient
MDL Number: MFCD07774120
CAS číslo: 42753-67-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR