3-Carboxymethylmorpholine-4-carboxylic acid tert-butyl ester 97%
Dodavatel: Apollo Scientific
Synonyms:
N-Boc-3-morpholineacetic acid, 4-Boc-3-Carboxymethylmorpholine
OR964953-1G
OR964953-250MG
APOSOR964953-1GEA
10650
CZK
APOSOR964953-1G
APOSOR964953-250MG
3-Carboxymethylmorpholine-4-carboxylic acid tert-butyl ester 97%
3-Carboxymethylmorpholine-4-carboxylic acid tert-butyl ester
Vzorec:
C₁₁H₁₉NO₅ MW: 245.28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04114891 CAS číslo: 859155-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...