Tisk…

N-Methyl-2-thiazolamine 97%

Dodavatel: Apollo Scientific

OR964710-1G OR964710-250MG OR964710-5G
APOSOR964710-1GEA 7670 CZK
APOSOR964710-1G APOSOR964710-250MG APOSOR964710-5G
N-Methyl-2-thiazolamine 97%
N-Methyl-2-thiazolamine
Vzorec: C₄H₆N₂S
Storage Temperature: Ambient
MDL Number: MFCD09966061
CAS číslo: 6142-06-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR