5-Methyl-1H-pyrrolo[2,3-b]pyridine 98%
Dodavatel: Apollo Scientific
OR964572-1G
APOSOR964572-1GEA
1700
CZK
APOSOR964572-1G
5-Methyl-1H-pyrrolo[2,3-b]pyridine 98%
5-Methyl-1H-pyrrolo[2,3-b]pyridine
Vzorec:
C8H8N2 MW: 132,1625 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06659674 CAS číslo: 824-52-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...