Boc-D-Glu-OMe
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(R)-4-((tert-Butoxycarbonyl)amino)-5-methoxy-5-oxopentanoic acid
OR964508-1G
OR964508-5G
APOSOR964508-1GEA
4370
CZK
APOSOR964508-1G
APOSOR964508-5G
Boc-D-Glu-OMe
Boc-D-Glu-OMe
Vzorec:
C₁₁H₁₉NO₆ MW: 261,274 g/mol Bod tání: 59…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00076930 CAS číslo: 55227-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...