Tisk…

(R)-2-Hydroxy-4-methylpentanoic acid 97%

Dodavatel: Apollo Scientific

Synonyms: D-α-Hydroxyisocaproic acid, (R)-Leucic acid

OR964438-5G
APOSOR964438-5GEA 3940 CZK
APOSOR964438-5G
(R)-2-Hydroxy-4-methylpentanoic acid 97%
(R)-2-Hydroxy-4-methylpentanoic acid
Vzorec: C₆H₁₂O₃
MW: 132,159 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00211263
CAS číslo: 20312-37-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR