Tisk…

Boc-N-Me-Nle-OH 97%

Dodavatel: Apollo Scientific

Synonyms: Boc-N-methyl-L-norleucine

OR964309-1G OR964309-5G
APOSOR964309-1GEA 3910 CZK
APOSOR964309-1G APOSOR964309-5G
Boc-N-Me-Nle-OH 97%
Boc-N-Me-Nle-OH
Vzorec: C₁₂H₂₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00153391
CAS číslo: 117903-25-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR