Tisk…

3-(3-Methoxyphenyl)propionic acid 97%

Dodavatel: Apollo Scientific

Synonyms: 3-(3-Methoxyphenyl)propanoic acid

OR964229-25G
APOSOR964229-25GEA 3150 CZK
APOSOR964229-25G
3-(3-Methoxyphenyl)propionic acid 97%
3-(3-Methoxyphenyl)propionic acid
Vzorec: C₁₀H₁₂O₃
MW: 180,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014027
CAS číslo: 10516-71-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR