Tisk…

H-D-Lys(Z)-OH 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: H-D-Lys(Cbz)-OH, Nepsilon-Benzyloxycarbonyl-D-lysine, Nepsilon-Cbz-D-lysine, H-D-Lys(Z)-OH, Nepsilon-Carbobenzoxy-D-lysine

OR964205-100G OR964205-25G
APOSOR964205-100GEA 11380 CZK
APOSOR964205-100G APOSOR964205-25G
H-D-Lys(Z)-OH 97%
H-D-Lys(Z)-OH
Vzorec: C₁₄H₂₀N₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00063120
CAS číslo: 34404-32-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR