Tisk…

Boc-D-norvaline 97%

Dodavatel: Apollo Scientific

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)pentanoic acid, Boc-D-Nva-OH

OR964138-25G OR964138-5G
APOSOR964138-25GEA 2440 CZK
APOSOR964138-25G APOSOR964138-5G
Boc-D-norvaline 97%
Boc-D-norvaline
Vzorec: C₁₀H₁₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD00155638
CAS číslo: 57521-85-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR