Tisk…

H-DL-Phe(3-CN)-OH 99%

Dodavatel: Apollo Scientific

Synonyms: 2-Amino-3-(3-cyanophenyl)propanoic acid

OR964107-1G OR964107-5G
APOSOR964107-1GEA 3190 CZK
APOSOR964107-1G APOSOR964107-5G
H-DL-Phe(3-CN)-OH 99%
H-DL-Phe(3-CN)-OH
Vzorec: C₁₀H₁₀N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00190859
CAS číslo: 63999-80-4
UN: 3439
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR