Ac-D-Nle-OH, Acetyl-D-2-aminohexanoic acid 95%
Dodavatel: Apollo Scientific
Synonyms:
Ac-d-nle-oh
OR964093-25G
OR964093-5G
APOSOR964093-25GEA
20900
CZK
APOSOR964093-25G
APOSOR964093-5G
Ac-D-Nle-OH, Acetyl-D-2-aminohexanoic acid 95%
Ac-D-Nle-OH, Acetyl-D-2-aminohexanoic acid
Vzorec:
C₈H₁₅NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00063171 CAS číslo: 54896-21-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...