(R)-(-)-Methyl mandelate 98%
Dodavatel: Apollo Scientific
Synonyms:
D(-)-Methyl glycolate, (R)-(-)-Methyl glycolate, (R)-(-)-Mandelic acid methyl ester, D(-)-Methyl mandelate, D(-)-2-Hydroxy-2-phenylacetic acid methyl ester, D(-)-Mandelic acid methyl ester, (R)-Methyl 2-hydroxy-2-phenylacetate, Methyl (R)-(-)-mandelate, (R)-(-)-2-Hydroxy-2-phenylacetic acid methyl ester
OR964083-100G
APOSOR964083-100GEA
2400
CZK
APOSOR964083-100G
(R)-(-)-Methyl mandelate 98%
(R)-(-)-Methyl mandelate
Vzorec:
C₉H₁₀O₃ MW: 166,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064247 CAS číslo: 20698-91-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...