Tisk…

N-(tert-Butoxycarbonyl)-D-threonine 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: N-Boc-D-threonine, Boc-D-Thr-OH, (2R,3S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxybutanoic acid

OR963919-100G OR963919-25G
APOSOR963919-100GEA 20700 CZK
APOSOR963919-100G APOSOR963919-25G
N-(tert-Butoxycarbonyl)-D-threonine 97%
N-(tert-Butoxycarbonyl)-D-threonine
Vzorec: C₉H₁₇NO₅
MW: 219,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037807
CAS číslo: 55674-67-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR