Tisk…

2-Chloro-6-(1-piperidinyl)pyrazine 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Chloro-6-(piperidin-1-yl)pyrazine

OR963879-1G OR963879-250MG
APOSOR963879-1GEA 5590 CZK
APOSOR963879-1G APOSOR963879-250MG
2-Chloro-6-(1-piperidinyl)pyrazine 95%
2-Chloro-6-(1-piperidinyl)pyrazine
Vzorec: C₉H₁₂ClN₃
MW: 197,67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864952
CAS číslo: 343856-62-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR