2-Bromo-5-phenyloxazole 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR963800-1G
OR963800-250MG
OR963800-5G
APOSOR963800-1GEA
9690
CZK
APOSOR963800-1G
APOSOR963800-250MG
APOSOR963800-5G
2-Bromo-5-phenyloxazole 96%
2-Bromo-5-phenyloxazole
Vzorec:
C₉H₆BrNO MW: 224,0540 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD12828094 CAS číslo: 129053-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...