Tisk…
(S)-Methyl-2-aminopentanoate hydrochloride 98%
Dodavatel: Apollo Scientific
Synonyms: H-Nva-OMe·HCl, H-Nva-Ome.HCl, L-Norvaline methyl ester hydrochloride
OR963720-100G
APOSOR963720-100GEA 3380 CZK
APOSOR963720-100G
(S)-Methyl-2-aminopentanoate hydrochloride 98%
(S)-Methyl-2-aminopentanoate hydrochloride
Vzorec: C₆H₁₄ClNO₂
MW: 167,633 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00190974
CAS číslo: 56558-30-6