Tisk…

2-Nitro-5-(1-pyrrolidinyl)aniline

Dodavatel: Apollo Scientific

Synonyms: 2-Nitro-5-(pyrrolidin-1-yl)aniline

OR963708-1G OR963708-5G
APOSOR963708-1GEA 2690 CZK
APOSOR963708-1G APOSOR963708-5G
2-Nitro-5-(1-pyrrolidinyl)aniline
2-Nitro-5-(1-pyrrolidinyl)aniline
Vzorec: C₁₀H₁₃N₃O₂
MW: 207.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00545947
CAS číslo: 289913-98-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR