Tisk…

2'-Amino-4'-methoxyacetophenone 95%

Dodavatel: Apollo Scientific

Synonyms: 1-(2-Amino-4-methoxyphenyl)ethanone

OR963707-1G OR963707-250MG
APOSOR963707-1GEA 4630 CZK
APOSOR963707-1G APOSOR963707-250MG
2'-Amino-4'-methoxyacetophenone 95%
2'-Amino-4'-methoxyacetophenone
Vzorec: C₉H₁₁NO₂
MW: 165.1900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972112
CAS číslo: 42465-53-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR