Tisk…

Boc-L-alaninal 95%

Dodavatel: Apollo Scientific

Synonyms: N-tert-Butoxycarbonyl-L-alaninal, Boc-L-Ala-aldehyde, Boc-L-alaninal, Boc-Ala-aldehyde, (S)-(1-Oxopropan-2-yl)carbamic acid tert-Butyl ester, (S)-tert-Butyl (1-Oxopropan-2-yl)carbamate

OR963550-5G
APOSOR963550-5GEA 4530 CZK
APOSOR963550-5G
Boc-L-alaninal 95%
Boc-L-alaninal
Vzorec: C₈H₁₅NO₃
MW: 173,212 g/mol
Bod tání: 82…84 °C
Storage Temperature: Freezer
MDL Number: MFCD00143786
CAS číslo: 79069-50-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR