2,3-Dihydro-1H-indole-7-carbonitrile 97%
Dodavatel: Apollo Scientific
OR963530-1G
OR963530-250MG
APOSOR963530-1GEA
6750
CZK
APOSOR963530-1G
APOSOR963530-250MG
2,3-Dihydro-1H-indole-7-carbonitrile 97%
2,3-Dihydro-1H-indole-7-carbonitrile
Vzorec:
C₉H₈N₂ Storage Temperature: Ambient |
MDL Number:
MFCD01830669 CAS číslo: 115661-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...