Tisk…

4-(4-Aminophenyl)benzonitrile 98%

Dodavatel: Apollo Scientific

OR963407-1G OR963407-5G
APOSOR963407-1GEA 1780 CZK
APOSOR963407-1G APOSOR963407-5G
4-(4-Aminophenyl)benzonitrile 98%
4-(4-Aminophenyl)benzonitrile
Vzorec: C₁₃H₁₀N₂
Storage Temperature: Ambient
MDL Number: MFCD00191367
CAS číslo: 4854-84-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR