Tisk…

N-Acetyl-D-valine 98%

Dodavatel: Apollo Scientific

Synonyms: Ac-D-Val-OH, N-Acetyl-D-Valine, N-Ac-D-Val-OH

OR963206-100G
APOSOR963206-100GEA 3540 CZK
APOSOR963206-100G
N-Acetyl-D-valine 98%
N-Acetyl-D-valine
Vzorec: C₇H₁₃NO₃
MW: 159,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00026457
CAS číslo: 17916-88-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR