D-Methionine methyl ester hydrochloride 96%
Dodavatel: Apollo Scientific
Synonyms:
H-D-Met-OMe·HCl, H-D-Met-OMe.HCl
OR963120-100G
OR963120-25G
APOSOR963120-100GEA
8010
CZK
APOSOR963120-100G
APOSOR963120-25G
D-Methionine methyl ester hydrochloride 96%
D-Methionine methyl ester hydrochloride
Vzorec:
C₆H₁₃NO₂S·HCl Storage Temperature: Ambient |
MDL Number:
MFCD00070385 CAS číslo: 69630-60-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...