Tisk…

N-Me-D-Ala-OtBu·HCl 98%

Dodavatel: Apollo Scientific

OR962965-1G OR962965-5G
APOSOR962965-1GEA 2900 CZK
APOSOR962965-1G APOSOR962965-5G
N-Me-D-Ala-OtBu·HCl 98%
N-Me-D-Ala-OtBu·HCl
Vzorec: C₈H₁₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD08275767
CAS číslo: 405513-14-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR