Tisk…

2-Methoxy-4-(4-methylpiperazin-1-yl)aniline 97%

Dodavatel: Apollo Scientific

OR962870-5G
APOSOR962870-5GEA 2150 CZK
APOSOR962870-5G
2-Methoxy-4-(4-methylpiperazin-1-yl)aniline 97%
2-Methoxy-4-(4-methylpiperazin-1-yl)aniline
Vzorec: C₁₂H₁₉N₃O
MW: 221,2988 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848954
CAS číslo: 122833-04-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR